SeCure® 'Y' Connector Kits, Restek
Supplier: Restek
20276 20277 20278
RK20276EA 431.17 USD
RK20276 RK20277 RK20278
SeCure® 'Y' Connector Kits, Restek
Fittings and Connectors
Combine the simplicity of a “Y” Press-Tight® connector with the strength of a metal union.
  • Reliable seal integrity
  • Open design allows visual confirmation of the seal
  • Use to connect two analytical columns to a transfer line or guard column
The ferrules and knurled nuts hold the fused silica tubing in place, which prevents the tubing from unexpectedly disconnecting, even at temperatures as high as 400°C.
Información para pedidos: Kits include: SeCure® “Y” connector body, three knurled nuts, universal “Y” Press-Tight® union, and three ferrules.