Vespel®/Graphite Ferrule ¹/₃₂", Restek
Supplier: Restek
20259 21039
RK20259PK 138.55 USD
RK20259 10852-588
Vespel®/Graphite Ferrule ¹/₃₂", Restek
Fittings and Connectors
Vespel®/Graphite ferrules are sized to fit 1/32" fittings.
Vespel®/Graphite hybrid material for excellent combination of sealing and workability.