Replacement CarboFrit® Inserts, Restek
Supplier: Restek
20295 20294
RK20295PK 97.37 USD
RK20295 75778-118
Replacement CarboFrit® Inserts, Restek
Chromatography Supplies Chromatography Spare Parts
Replacement frits are used for liners with internal diameters of ≤4 mm.