Capillary Ferrules for ¹/₁₆" Compression-Type Fittings, Restek
Supplier: Restek
20275 27061
RK20233PK 51.45 USD
RK20233 10057-900 10852-506 10852-602
Capillary Ferrules for ¹/₁₆" Compression-Type Fittings, Restek
Fittings and Connectors
Capillary ferrules are sized to fit 1/16" fittings.
  • Available in Vespel®, graphite, or Vespel®/graphite material
For micropacked columns or 0.28 mm MXT columns.
Información para pedidos: For micropacked columns or 0.28 mm MXT columns.