Forma™ Environmental Chambers, Thermo Scientific
Supplier: Thermo Fisher Scientific
3949 3920 3911 3940
101623-078EA 40307.69 USD
101623-078 101623-152 101623-100 101623-076
Forma™ Environmental Chambers, Thermo Scientific
Testing Chambers Environmental Test Chambers
Forma™ environmental chambers, which can be configured for a variety of purposes and are available in multiple capacities.
  • Wide temperature range of 0° to 60°C that enhances the flexibility of the unit
  • Relative humidity control via steam generator from above ambient to 95% at 37°C
  • Sophisticated digital controls
  • Programmable ramp and soak cycles for temperature and RH
  • Durable cabinets with true laminar airflow through side plenums for extremely tight uniformity
  • Phenolic-coated evaporator for use in applications that create corrosive byproducts
  • Manual control of defrost and refrigeration cycles
Cold-rolled steel, powder-coated cabinet with 2″ (5.1cm) thick, CFC-free and foamed-in-place polyurethane insulation. Stainless-steel interior with solid stainless-steel adjustable shelves and directed horizontal airflow system for excellent uniformity.