STERIL-PEEL™ Self-Seal Sterilization Rolls, GS Medical Packaging
Supplier: GS Medical Packaging
221439 221440 221432 221433 221434 221436
89201-540CS 556.66 USD
89201-540 89201-544 89201-542 89201-538 89201-536 89201-546
STERIL-PEEL™ Self-Seal Sterilization Rolls, GS Medical Packaging
Sterilization Indicators
All STERIL-PEEL™ products are manufactured from medical grade bleached kraft paper
STERIL-PEEL™ products are printed with dual indicators which change color in either steam or ethylene oxide. Both indicators conform with the latest ISO 11140 and EN868 regulations. The steam indicator ink will only change in the presence of all necessary parameters (saturated steam, temperature, and time).
Hydrogen peroxide, rather than chlorine, is used as the bleaching agent. They are made from virgin pulp, ensuring optimum consistency of the paper sheet, with superior barrier properties. All rolls are lead-free and feature medically safe inks.
Certificaciones: 510K and CE compliant.