Uric Acid Reagent Set, Pointe Scientific
Supplier: HORIBA INSTRUMENTS INCORPORATED
U7581-500 U7581-1L
10022-970EA 303.35 USD
10022-970 10022-972
Uric Acid Reagent Set, Pointe Scientific
Assays Enzyme Assays
Uric Acid Reagent Set is used for the quantitative determination of Uric Acid in serum.
  • Quantitative determination of Uric Acid in serum
  • For in vitro diagnostic use only
  • Absorbance at 520nm is proportional to the concentration of Uric Acid in the sample
Uric Acid Reagent Set is used for the quantitative determination of Uric Acid in serum. For in vitro diagnostic use only. Uric Acid is oxidized by Uricase to allantoin and hydrogen peroxide. HDCBS + 4-AAP + hydrogen peroxide, in the presence of peroxidase, produces a red chromagen that is measured at 520nm. The absorbance at 520nm is proportional to the concentration of Uric Acid in the sample.