Buffer solutions, color-coded, Thermo Scientific

Supplier: Thermo Scientific Chemicals

61104-0040 61106-0040
200057-264EA 57.51 USD
200057-264 200055-280 200057-268 200057-272 TS61104-0040 TS61106-0040
Buffer solutions, color-coded, Thermo Scientific
Buffers pH Calibration Buffers

Storage Temperature: Ambient


  • Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR