Methylenediphenyl-4,4'-diisocyanate 98%, soluble

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4,4'-Diphenylmethane Diisocyanate, Methylene di-p-phenyl diisocyanate, Diphenylmethane 4,4'-diisocyanate, Bis(4-isocyanatophenyl)methane, MDI, 4,4'-Methylenebis(phenyl Isocyanate)

AC41428-1000 AC41428-5000
200058-050EA 27.14 USD
200058-050 200058-052
Methylenediphenyl-4,4'-diisocyanate 98%, soluble
Methylenediphenyl-4,4'-diisocyanate

Appearance: White to yellow Flakes

Formula: C₁₅H₁₀N₂O₂
MW: 250.26 g/mol
Boiling Pt: 392 °C (760 mmHg)
Melting Pt: 40 °C
Density: 1.22 g/cm³
Flash Pt: 202 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036131
CAS Number: 101-68-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR