DL-1-(2-Furyl)ethanol 99% stabilised
Supplier: Thermo Scientific Chemicals
|
Warning
|
AC33967-0050
AC33967-0010
200006-374EA
81.42
USD
200006-374
200043-358
DL-1-(2-Furyl)ethanol 99% stabilised
DL-1-(2-Furyl)ethanol
Appearance: Clear colorless to light yellow Liquid
Formula:
C₆H₈O₂ MW: 112.128 g/mol Boiling Pt: 72…74 °C (15 mmHg) Density: 1.07 g/cm³ Storage Temperature: Refrigerator |
MDL Number:
MFCD00077775 CAS Number: 4208-64-4 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.4770 to 1.4800 (20 °C, 589 nm) |
HPLC | ≥98.5 % |
Stabilizer | 0.3 to 0.5 % hydroquinone |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...