Anti-IgA Goat Polyclonal Antibody (CF™ 594)

Supplier: Biotium

20429-500UL 20429-50UL
10018-356EA 217.08 USD
10018-356 10018-358
Anti-IgA Goat Polyclonal Antibody (CF™ 594)
Antibodies
Goat anti-human IgA reacts specifically with IgA heavy chains (α chains) and not with immunoglobulin light chains. To minimize crossreactivity, it has been adsorbed human IgG prior to conjugation. This conjugate is labeled with CF™594 dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Deep-red fluorescent CF™594 (Ex/Em 593/614 nm) is spectrally similar to Alexa Fluor® 594 and Texas Red® dye but yields much brighter protein conjugate, and has excellent photostability.

Alexa Fluor® and Texas Red® are registered trademarks of Thermo Fisher Scientific.

Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: CF594
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR