Anti-IgG Lama Polyclonal Antibody (CF™ 594)

Supplier: Biotium

20451-500UL 20451-50UL
10001-918EA 228.45 USD
10001-918 10001-920
Anti-IgG Lama Polyclonal Antibody (CF™ 594)
Antibodies
Llama anti-rabbit CF™ dye conjugates are prepared by labeling affinity purified llama anti-rabbit IgG (H+L) with a selection of CF™ dyes. CF™ dyes offer combined advantages in brightness, photostability, and specificity compared other fluorescent dyes. CF™594 (Ex/Em 593/614 nm) is a deep-red dye spectrally similar to Alexa Fluor® 594 and Texas Red® dye but yields much brighter protein conjugates due to its high quantum yield and exceptional water solubility. CF™594 also has excellent photostability ideal for demanding applications, such as confocal microscopy and single molecule imaging.

Alexa Fluor® and Texas Red® are registered trademarks of Thermo Fisher Scientific.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF594
Epitope:
Host: Llama
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR