Anti-IgG gamma Goat Polyclonal Antibody (CF™ 647)

Supplier: Biotium

20448-500UL 20448-50UL
10001-900EA 266.7 USD
10001-900 10001-902
Anti-IgG gamma Goat Polyclonal Antibody (CF™ 647)
Antibodies
Goat anti-human IgG, Fc gamma specific antibody reacts specifically with the human IgG heavy (γ) chains and not with light chains and F(ab’)2 fragment on all human immunoglobulins. To minimize cross-reactivity, it has been adsorbed against bovine, horse, mouse and rabbit serum proteins prior to conjugation. Far-red fluorescent CF™647 (Ex/Em 650/665 nm) is brighter than Cy®5 and an excellent alternative to Alexa Fluor® 647 for antibody conjugates.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF647
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR