Anti-IgG gamma Goat Polyclonal Antibody (CF™ 594)

Supplier: Biotium

20446-500UL 20446-50UL
10001-888EA 266.7 USD
10001-888 10001-890
Anti-IgG gamma Goat Polyclonal Antibody (CF™ 594)
Antibodies
Goat anti-human IgG, Fc gamma specific antibody reacts specifically with the human IgG heavy (γ) chains and not with light chains and F(ab’)2 fragment on all human immunoglobulins. To minimize cross-reactivity, it has been adsorbed against bovine, horse, mouse and rabbit serum proteins prior to conjugation. CF™594 (Ex/Em 593/614 nm) is a deep-red dye spectrally similar to Alexa Fluor® 594 and Texas Red® dye but yields much brighter protein conjugates due to its high quantum yield and exceptional water solubility. CF™594 also has excellent photostability ideal for demanding applications, such as confocal microscopy and single molecule imaging.

Alexa Fluor® and Texas Red® are registered trademarks of Thermo Fisher Scientific.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF594
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR