Capreomycin sulphate 90%

Supplier: Thermo Scientific Chemicals
Danger

J66684-03 J66684-MD
10067-248EA 142 USD
10067-248 AAJ66684-MD
Capreomycin sulphate 90%
Capreomycin sulphate

Thought to inhibit protein synthesis by inhibiting normal ribosomal functions


White powder


Freely soluble in water


Precaución: Research use only: Not intended for animal or human diagnostic or therapeutic use.

Formula: C₂₅H₄₄N₁₄O₈·H₂SO₄
Boiling Pt: 1377 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Freezer
MDL Number: MFCD00079032
CAS Number: 1405-37-4
EINECS: 215-776-8

Order Now

Specification Test Results

Assay 90%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR