ASI Cytomegalovirus, Arlington Scientific
Supplier: ARLINGTON SCIENTIFIC INC.
820096AG 820096AM
10853-064KT 444.85 USD
10853-064 10853-066
ASI Cytomegalovirus, Arlington Scientific
Assays ELISAs
ELISA Kit detects human antibodies to Cytomegalovirus.
  • Qualitative and semiquantitative detection
  • Detects human IGG or IGM antibodies
ELISA for the qualitative and semiquantitative detection of human IGG/IGM antibodies to Cytomegalovirus.
Envase: Kit of 96