Vasculife® SMC Medium, Lifeline Cell Technology
Supplier: Lifeline Cell Technology
LL-0014 LS-1040
102970-820EA 219.23 USD
102970-820 102970-856
Vasculife® SMC Medium, Lifeline Cell Technology
Cell Culture Media
VascuLife® SMC Smooth Muscle Cell Medium is a Lifeline® low-serum medium optimized for the culture of human smooth muscle cells.
  • Optimized for the culture of human smooth muscle cells
  • Contains no antimicrobials and no phenol red
  • Prepare fresh medium
VascuLife® SMC Smooth Muscle Cell Medium is a Lifeline® low-serum medium optimized for the culture of human smooth muscle cells (SMC). VascuLife® SMC supports the growth of these cells over a period of at least 15 population doublings. VascuLife® Basal Medium contains no antimicrobials and no phenol red, since these components can affect results.
The VascuLife® SMC Medium Complete Kit includes VascuLife® Basal Medium and VascuLife® SMC LifeFactors® so you can prepare fresh medium.
Información para pedidos: The VascuLife® SMC LifeFactors® Kit includes: rh FGF basic 0.5 mL, rh Insulin 0.5 mL, Ascorbic Acid 0.5 mL, L-Glutamine 25 mL, rh EGF 0.5 mL, FBS 25 mL, Gentamicin and Amphotericin B 0.5 mL.
VascuLife® SMC Medium Complete Kit contains VascuLife® Basal Medium and the VascuLife® LifeFactors® Kit. VascuLife® Basal Medium requires the appropriate LifeFactors®.