Alcohol Assay Kits, Cell Biolabs
Supplier: Cell Biolabs
STA-620 STA-621
102965-242EA 578.46 USD
102965-242 102965-244
Alcohol Assay Kits, Cell Biolabs
Assays Enzyme Assays
Cell Biolabs’ Alcohol Assay Kits measure primary alcohols by an enzymatic, oxidation reaction, producing hydrogen peroxide which reacts with the kit’s probe
These assay kits are available with either colorimetric or fluorometric detection. The colorimetric assay has a detection sensitivity limit of ~30 µM (0.0001 % w/v) and the fluorometric assay has a detection sensitivity limit of ~15 µM (0.00007 % w/v). They can detect various primary alcohols and are not ethanol specific. Each kit provides sufficient reagents to perform up to 100 assays, including blanks, standards and unknown samples.