Free Fatty Acid Assay Kits, Cell Biolabs
Supplier: Cell Biolabs
STA-618
STA-619
102965-238EA
596.92
USD
102965-238
102965-240
Free Fatty Acid Assay Kits, Cell Biolabs
Assays
Enzyme Assays
Cell Biolabs' Free Fatty Acid Assay Kits contain a palmitic acid standard and is a simple assay that quantitatively measures the non-esterified free fatty acid (NEFA) concentration in various samples using a 96-well microtiter plate format
Each kit provides sufficient reagents to perform up to 100 assays, including blanks, standards and unknown samples. Kits are available for both colorimetric and fluorometric detection methods.