RenaLife™ Epithelial Medium
Supplier: Lifeline Cell Technology
LL-0025 LS-1048 LM-0010
102970-824EA 223.23 USD
102970-824 102970-862 102970-948
RenaLife™ Epithelial Medium
Cell Culture Media
RenaLife™ Renal Epithelial Medium is specifically optimized for the low-serum culture of human renal epithelial cells.
  • Supports superior proliferation at different seeding densities
  • Contains no phenol red or antimicrobials
  • optimized for the low-serum culture of human renal epithelial cells
In comparisons with other commercially available media, RenaLife™ supports superior proliferation at different seeding densities. RenaLife™ Basal Medium contains no phenol red or antimicrobials. RenaLife™ is optimized for the low-serum culture of human renal epithelial cells over a period of at least 15 population doublings.
The LifeFactors® Kit includes: rh Insulin 0.5 mL, Epinephrine 0.5 mL, Hydrocortisone Hemisuccinate 0.5 mL, FBS 2.5 mL, Triiodothyronine 0.5 mL, rh EGF 1 mL, Transferrin PS 0.5 mL, L-Alanyl L-Glutamine 0.5 mL, Gentamicin and Amphotericin B 0.5 mL.
Información para pedidos: RenaLife™ Medium Complete Kit includes RenaLife™ Basal Medium and RenaLife™ LifeFactors® Kit. RenaLife™ Basal Medium requires the appropriate LifeFactors®.