Sklar Blue™ Graves OB-View Vaginal Speculum, OR Grade, Sklar®
Supplier: Sklar
91-3740 91-3741
10184-314EA 496.42 USD
10184-314 10184-316
Sklar Blue™ Graves OB-View Vaginal Speculum, OR Grade, Sklar®
Specula
The Sklar Blue™ Graves OB-View Vaginal Speculum is used for electrocsurgical gynecological procedures, such as LEEP.
  • Sklar Blue™ instruments are fully autoclavable and may also be cleaned ultrasonically
It is a standard double blade duckbill speculum that features a wide proximal opening for instrumentation. The blades are then opened by an external lever and screw. The Sklar Blue™ Graves Vaginal Speculum is available with a permanent smoke evacuation tube, a loop for disposable smoke evacuation tubing, or no smoke evacuation. Sklar Blue™ electrosurgical instruments feature high quality stainless steel with an insulated coating. This safe coating prevents radio frequency currents from being transmitted through the instrument which reduces the burning of non-targeted tissue.
The Graves Speculum has wider blades than the Pederson Speculum and the practitioner can best determine which type and size speculum is most appropriate for the patient.
Certificaciones: Comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79.