Sklarlite XD™ Adson Tissue Forceps, OR Grade, Sklar®
Supplier: Sklar
23-2606 23-2608
10195-346EA 130.53 USD
10195-346 10195-348
Sklarlite XD™ Adson Tissue Forceps, OR Grade, Sklar®
Forceps
Sklarlite XD™ Adson Tissue Forceps are for holding and manipulating delicate tissues
These forceps have a wide, flat thumb grasp area that is commonly serrated. The jaws are short and the tips are narrow. Sklarlite XD™ instruments are noticeably lighter and delicate than standard instruments which allow the surgeon the highest degree of precision with the least amount of fatigue. These instruments have been uniquely manufactured to be one of the lightest, most delicate general instruments available today. This product is straight with smooth jaws, 1 X 2 teeth, and a length of 4-3/4 inches.