Rapid RCA Assay Kit, Cell Biolabs
Supplier: Cell Biolabs
VPK-111 VPK-111-5
102965-352EA 596.92 USD
102965-352 102965-354
Rapid RCA Assay Kit, Cell Biolabs
Assays Cellular Assays
Cell Biolabs' Rapid RCA Assay Kit uses an immunocytochemistry staining protocol that requires only a two day incubation
Traditionally RCA (replication competent adenovirus) is measured in permissive cells by a plaque-forming unit (PFU) assay which takes 10-14 days. This assay is faster and more accurate than traditional plaque-forming unit assays and requires no agar overlay steps