OxiSelect™ Human Oxidized HDL ELISA Kits, Cell Biolabs
Supplier: Cell Biolabs
STA-869 STA-888 STA-889
102965-326EA 873.85 USD
102965-326 102965-330 102965-332
OxiSelect™ Human Oxidized HDL ELISA Kits, Cell Biolabs
Assays ELISAs
OxiSelect™ Human Oxidized HDL ELISA Kits provide a convenient method to measure one of the markers of Oxidized HDL (OxHDL) in human serum or plasma samples
Kits are available to measure one of three markers of oxidation on the protein portion of HDL: MDA-HDL, CML-HDL, or HNE-HDL. The kits contain a standard against which unknown samples may be measured