OxiSelect™ Human Oxidized LDL ELISA Kits, Cell Biolabs
Supplier: Cell Biolabs
STA-358 STA-369 STA-388 STA-389
102965-000EA 824.62 USD
102965-000 102965-040 102965-064 102965-066
OxiSelect™ Human Oxidized LDL ELISA Kits, Cell Biolabs
Assays ELISAs
OxiSelect™ Human Oxidized LDL ELISA Kits provide a convenient method to measure one of the markers of Oxidized LDL (OxLDL) in human serum or plasma samples
Kits are available to measure one of three markers of oxidation on the protein portion of LDL: MDA-LDL, CML-LDL, or HNE-LDL. In addition, the OxPL kit measures oxidation of the phospholipid portion of LDL. The kits contain a copper oxidized LDL as a standard against which unknown samples may be measured.