Glass-Klenz® Glassware Detergentz, STERIS®
Supplier: Steris
Glass-Klenz®
111408
21899-881CS 439.92 USD
21899-881
Glass-Klenz® Glassware Detergentz, STERIS®
Glass-Klenz® Glassware detergent is a low-foaming, chelant-based, liquid detergent for cleaning laboratory glasswares.
  • Effective removal of laboratory residues in both soft and hard water
  • Effectively holds soils in suspension allowing to be completely removed without redeposition
  • Reduces utility consumption and cleaning time due
  • Mild pH for enhanced safety and substrate compatibility
It is a free rinsing detergent specially formulated for use in automated glassware washing applications for the removal of particulate.
Certificaciones: Manufactured in a cGMP compliant, FDA registered and ISO 9001 certified facility.