Anti-IFN-g Mouse Monoclonal Antibody (Brilliant Violet® 785) [clone: 4S.B3]

Supplier: Biolegend
502541 502542
502541-BLEA 272.7 USD
502541-BL 10019-722
Anti-IFN-g Mouse Monoclonal Antibody (Brilliant Violet® 785) [clone: 4S.B3]
Antibodies
Brilliant Violet 785™ anti-human IFN-γ [4S.B3]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IFN-g
Clonality: Monoclonal
Clone: 4S.B3
Conjugation: Brilliant Violet® 785
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee;Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR