Anti-Granzyme B Mouse Monoclonal Antibody [clone: O94E6/GZMB]

Supplier: Biolegend
674301 674302
10020-022EA 113.7 USD
10020-022 10020-024
Anti-Granzyme B Mouse Monoclonal Antibody [clone: O94E6/GZMB]
Antibodies
Purified anti-Granzyme B [O94E6/GZMB]; Isotype: Mouse IgG2a; Reactivity: Human; Apps: WB; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.2 - 1.0 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Granzyme B
Clonality: Monoclonal
Clone: O94E6/GZMB
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR