Anti-Cutaneous Lymphocyte Antigen (CLA) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: HECA-452]

Supplier: Biolegend
321311 321312
10019-284EA 123.9 USD
10019-284 10019-286
Anti-Cutaneous Lymphocyte Antigen (CLA) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: HECA-452]
Antibodies
PE anti-human/mouse Cutaneous Lymphocyte Antigen (CLA) [HECA-452]; Isotype: Rat IgM, κ; Reactivity: Human, Mouse; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Cutaneous Lymphocyte Antigen (CLA)
Clonality: Monoclonal
Clone: HECA-452
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: Rat IgM, κ
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR