Anti-CD56 Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: 5.1H11]

Supplier: Biolegend
362533 362534
10021-064EA 217.3 USD
10021-064 10021-066
Anti-CD56 Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: 5.1H11]
Antibodies
Brilliant Violet 510™ anti-human CD56 (NCAM) [5.1H11]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD56
Clonality: Monoclonal
Clone: 5.1H11
Conjugation: Brilliant Violet® 510
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR