Anti-CD319 (CRACC) Mouse Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: 162.1]

Supplier: Biolegend
331811 331812
10019-332EA 179.95 USD
10019-332 10019-334
Anti-CD319 (CRACC) Mouse Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: 162.1]
Antibodies
PE/Dazzle™ 594 anti-human CD319 (CRACC) [162.1]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cynomolgus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD319 (CRACC)
Clonality: Monoclonal
Clone: 162.1
Conjugation: PE (Phycoerythrin)/Dazzle 594
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR