Anti-CD303 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 201A]

Supplier: Biolegend
354211 354212
10759-730EA 242.2 USD
10759-730 10019-498
Anti-CD303 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: 201A]
Antibodies
Brilliant Violet 421™ anti-human CD303 (BDCA-2) [201A]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD303
Clonality: Monoclonal
Clone: 201A
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR