Anti-CD267 (TACI) Rat Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: 1A1]

Supplier: Biolegend
311909 311910
10019-248EA 179 USD
10019-248 10019-250
Anti-CD267 (TACI) Rat Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: 1A1]
Antibodies
PE/Dazzle™ 594 anti-human CD267 (TACI) [1A1]; Isotype: Rat IgG2a, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD267 (TACI)
Clonality: Monoclonal
Clone: 1A1
Conjugation: PE (Phycoerythrin)/Dazzle 594
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR