Anti-CD22 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: S-HCL-1]

Supplier: Biolegend
363503 363504
10019-526EA 131.7 USD
10019-526 10019-528
Anti-CD22 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: S-HCL-1]
Antibodies
PE anti-human CD22 [S-HCL-1]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD22
Clonality: Monoclonal
Clone: S-HCL-1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR