Anti-CD210 (IL-10 R) Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 3F9]

Supplier: Biolegend
308811 308812
10021-032EA 147.5 USD
10021-032 10021-034
Anti-CD210 (IL-10 R) Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 3F9]
Antibodies
APC anti-human CD210 (IL-10 R) [3F9]; Isotype: Rat IgG2a, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD210 (IL-10 R)
Clonality: Monoclonal
Clone: 3F9
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR