Anti-CD140a Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 16A1]

Supplier: Biolegend
323507 323508
10019-294EA 179 USD
10019-294 10019-296
Anti-CD140a Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 16A1]
Antibodies
PE/Cy7 anti-human CD140a (PDGFRα) [16A1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD140a
Clonality: Monoclonal
Clone: 16A1
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR