Anti-BrdU Mouse Monoclonal Antibody [clone: 3D4]

Supplier: Biolegend
364101 364102
10019-572EA 73.2 USD
10019-572 10019-574
Anti-BrdU Mouse Monoclonal Antibody [clone: 3D4]
Antibodies
Purified anti-BrdU [3D4]; Isotype: Mouse IgG1, κ; Apps: ICFC, IHC; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: BrdU
Clonality: Monoclonal
Clone: 3D4
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR