Anti-BrdU Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 3D4]

Supplier: Biolegend
364103 364104
10019-576EA 100.2 USD
10019-576 10019-578
Anti-BrdU Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 3D4]
Antibodies
FITC anti-BrdU [3D4]; Isotype: Mouse IgG1, κ; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: BrdU
Clonality: Monoclonal
Clone: 3D4
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR