Anti-IgM Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: MHM-88]

Supplier: Biolegend
314525 314526
10758-140EA 242.2 USD
10758-140 10758-142
Anti-IgM Mouse Monoclonal Antibody (Brilliant Violet® 650) [clone: MHM-88]
Antibodies
Brilliant Violet 650™ anti-human IgM [MHM-88]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IgM
Clonality: Monoclonal
Clone: MHM-88
Conjugation: Brilliant Violet® 650
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR