Anti-IgG2a, k isotype ctrl Mouse Monoclonal Antibody (Brilliant Violet® 711) [clone: MOPC-173]

Supplier: Biolegend
400271 400272
10761-108EA 229.8 USD
10761-108 10761-110
Anti-IgG2a, k isotype ctrl Mouse Monoclonal Antibody (Brilliant Violet® 711) [clone: MOPC-173]
Antibodies
Brilliant Violet 711™ Mouse IgG2a, κ Isotype Ctrl [MOPC-173]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse; Apps: FC, ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest. Use our Concentration Lookup tool to find the exact concentrations of your lots of product.

Type: Control
Antigen: IgG2a, k isotype ctrl
Clonality: Monoclonal
Clone: MOPC-173
Conjugation: Brilliant Violet® 711
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR