Anti-IgG2a, k Isotype Ctrl Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: RTK2758]

Supplier: Biolegend
400535 400536 400549
400535-BLEA 236.23 USD
400535-BL 400536-BL 10761-172
Anti-IgG2a, k Isotype Ctrl Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: RTK2758]
Antibodies
Brilliant Violet 421™ Rat IgG2a, κ Isotype Ctrl [RTK2758]; Isotype: Rat IgG2a, κ; Reactivity: KLH; Apps: FC, ICFC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis as negative control. Use at concentrations comparable to those of the specific antibody of interest.

Type: Control
Antigen: IgG2a, k Isotype Ctrl
Clonality: Monoclonal
Clone: RTK2758
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: KLH
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR