Anti-HLA-DR Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: Tü36]

Supplier: Biolegend
361609 361610
10760-976EA 167.1 USD
10760-976 10760-978
Anti-HLA-DR Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: Tü36]
Antibodies
APC anti-human HLA-DR [Tü36]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cross-Reactivity: Baboon, Cynomolgus, Cat (Feline), Dog (Canine), Cattle (Bovine, Cow); Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: HLA-DR
Clonality: Monoclonal
Clone: Tü36
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;Cross-Reactivity: Baboon;Cynomolgus;Cat (Feline);Dog (Canine);Cattle (Bovine;Cow)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR