Anti-Fc epsilon RI alpha Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: AER-37 (CRA-1)]

Supplier: Biolegend
334621 334622
334621-BLEA 192.5 USD
334621-BL 334622-BL
Anti-Fc epsilon RI alpha Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: AER-37 (CRA-1)]
Antibodies
PerCP/Cy5.5 anti-human FcεRIα [AER-37 (CRA-1)]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Fc epsilon RI alpha
Clonality: Monoclonal
Clone: AER-37 (CRA-1)
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR