Anti-CXCL9 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: J1015E10]

Supplier: Biolegend
357903 357904
10759-618EA 158.7 USD
10759-618 10759-620
Anti-CXCL9 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: J1015E10]
Antibodies
PE anti-human CXCL9 (MIG) [J1015E10]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CXCL9
Clonality: Monoclonal
Clone: J1015E10
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR