Anti-CD86 Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: IT2.2]

Supplier: Biolegend
305431 305432
10757-912EA 229.8 USD
10757-912 10757-914
Anti-CD86 Mouse Monoclonal Antibody (Brilliant Violet® 510) [clone: IT2.2]
Antibodies
Brilliant Violet 510™ anti-human CD86 [IT2.2]; Isotype: Mouse IgG2b, κ; Reactivity: Human, African Green, Baboon, Capuchin Monkey, Common Marmoset, Cotton-topped Tamarin, Chimpanzee, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: IT2.2
Conjugation: Brilliant Violet® 510
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;African Green;Baboon;Capuchin Monkey;Common Marmoset;Cotton-topped Tamarin;Chimpanzee;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR