Anti-CD74 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: LN2]

Supplier: Biolegend
326811 326812
10758-340EA 141.4 USD
10758-340 10758-342
Anti-CD74 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: LN2]
Antibodies
APC anti-human CD74 [LN2]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD74
Clonality: Monoclonal
Clone: LN2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR