Anti-CD56 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: 5.1H11]

Supplier: Biolegend
362519 362520
10760-884EA 153.1 USD
10760-884 10760-886
Anti-CD56 Mouse Monoclonal Antibody (PB (Pacific Blue)) [clone: 5.1H11]
Antibodies
Pacific Blue™ anti-human CD56 (NCAM) [5.1H11]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD56
Clonality: Monoclonal
Clone: 5.1H11
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR