Anti-CD5 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 53-7.3]

Supplier: Biolegend
100621 100622
10756-686EA 153.1 USD
10756-686 10756-688
Anti-CD5 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 53-7.3]
Antibodies
PE/Cy7 anti-mouse CD5 [53-7.3]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: 53-7.3
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR