Anti-CD45 Rat Monoclonal Antibody (Brilliant Violet® 605) [clone: 30-F11]

Supplier: Biolegend
103139 103140
10759-130EA 293 USD
10759-130 10759-132
Anti-CD45 Rat Monoclonal Antibody (Brilliant Violet® 605) [clone: 30-F11]
Antibodies
Brilliant Violet 605™ anti-mouse CD45 [30-F11]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 500 μl

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining with μg, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. For flow cytometric staining with μl, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: 30-F11
Conjugation: Brilliant Violet® 605
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR