Anti-CD4 Rat Monoclonal Antibody (Brilliant Violet® 711) [clone: RM4-5]

Supplier: Biolegend
100549 100550 100557
100549-BLEA 289.98 USD
100549-BL 10756-670 10759-036
Anti-CD4 Rat Monoclonal Antibody (Brilliant Violet® 711) [clone: RM4-5]
Antibodies
Brilliant Violet 711™ anti-mouse CD4 [RM4-5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For immunofluorescent staining using the μg size, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. For immunofluorescent staining using the μl size, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RM4-5
Conjugation: Brilliant Violet® 711
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR